4-(Iodomethyl)tetrahydro-2H-pyran 97%

Lieferant: Thermo Fisher Scientific
Gefahr

448700100
ACRO448700100EA 529 EUR
ACRO448700100
4-(Iodomethyl)tetrahydro-2H-pyran 97%
4-(Iodomethyl)tetrahydro-2H-pyran
Formel: C₆H₁₁IO
Lagertemperatur: Raumtemperatur
MDL: MFCD06410724
CAS-Nummer: 101691-94-5
UN: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR