2-Methyl-4-pyridinboronsäurepinakolester 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methylpyridine-4-boronic acid Pinacol ester, 2-(2-Methyl-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

448720010
ACRO448720010EA 410 EUR
ACRO448720010
2-Methyl-4-pyridinboronsäurepinakolester 95%
2-Methyl-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219,09 g/mol
Schmelzpunkt: 66…71 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08061590
CAS-Nummer: 660867-80-1

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 63 to 70 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR