1-Brom-5-chlor-2-iodbenzol 98%

Lieferant: Thermo Scientific
Achtung

448760250
ACRO448760250EA 292 EUR
ACRO448760250
1-Brom-5-chlor-2-iodbenzol 98%
1-Brom-5-chlor-2-iodbenzol
Formel: C₆H₃BrClI
Molecular Weight: 317,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672944
CAS-Nummer: 31928-44-6

Bestellen

Spezifikation Testergebnisse

Appearance Orange to brown Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR