6-Chloro-7-iodo-7-deazapurine 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine

448850010 448850050
ACRO448850010EA 109 EUR
ACRO448850010 ACRO448850050
6-Chloro-7-iodo-7-deazapurine 96%
6-Chloro-7-iodo-7-deazapurine
Formel: C₆H₃ClIN₃
Molecular Weight: 279.47 g/mol
Schmelzpunkt: 179…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09263258
CAS-Nummer: 123148-78-7

Bestellen

Spezifikation Testergebnisse

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 180 °C
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR