2,2-Difluor-1,3-benzodioxol-5-carbonsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,2-Difluorobenzo[d][1,3]dioxole-5-carboxylic acid

449140050
ACRO449140050EA 250 EUR
ACRO449140050
2,2-Difluor-1,3-benzodioxol-5-carbonsäure 97%
2,2-Difluor-1,3-benzodioxol-5-carbonsäure
Formel: C₈H₄F₂O₄
Molecular Weight: 202,12 g/mol
Schmelzpunkt: 153…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00792417
CAS-Nummer: 656-46-2

Bestellen

Spezifikation Testergebnisse

Appearance Light grey to tan Powder
Infrared spectrum Conforms
Melting point 154 to 157 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR