N-Boc-4-oxo-L-proline methyl ester 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(S)-1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, Boc-4-oxo-Pro-OMe
449270010
449270050
ACRO449270010EA
60.8
EUR
ACRO449270010
ACRO449270050
N-Boc-4-oxo-L-proline methyl ester 97%
N-Boc-4-oxo-L-proline methyl ester
Formel:
C₁₁H₁₇NO₅ Schmelzpunkt: 42…45 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01861778 CAS-Nummer: 102195-80-2 |
Spezifikation Testergebnisse
Appearance | White to tan Solid |
Infrared spectrum | Conforms |
Melting point | 42 to 46 °C |
GC | ≥96.0 % |
Specific optical rotation | +10° to +17° (20 °C, 589 nm) (c=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...