1-Acetylpyren 97%

Lieferant: Thermo Scientific

Synonyme: 1-(Pyren-1-yl)ethanone

449300050
ACRO449300050EA 94.9 EUR
ACRO449300050
1-Acetylpyren 97%
1-Acetylpyren
Formel: C₁₈H₁₂O
Molecular Weight: 244,29 g/mol
Schmelzpunkt: 86…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00021663
CAS-Nummer: 3264-21-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 86 to 89 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR