(1-Formylcyclopropyl)carbamic acid-tert-butyl ester 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: tert-Butyl 1-formylcyclopropylcarbamate, 1(tert-Butoxycarbonylamino)cyclopropanecarboxaldehyde

449950010
ACRO449950010EA 172 EUR
ACRO449950010
(1-Formylcyclopropyl)carbamic acid-tert-butyl ester 97%
(1-Formylcyclopropyl)carbamic acid-tert-butyl ester
Formel: C₉H₁₅NO₃
Molecular Weight: 185,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697899
CAS-Nummer: 107259-06-3
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 82 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR