4-Fluoro-2-methoxypyridine 97%
Lieferant: Thermo Scientific
|
Achtung
|
450010010
ACRO450010010EA
260
EUR
ACRO450010010
4-Fluoro-2-methoxypyridine 97%
4-Fluoro-2-methoxypyridine
Formel:
C₆H₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835267 CAS-Nummer: 96530-81-3 |
Spezifikation Testergebnisse
Appearance | Colorlesss to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4740 to 1.4760 (20°C, 589 nm) |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...