5-Boc-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate
450020010
ACRO450020010EA
369
EUR
ACRO450020010
5-Boc-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine 97%
5-Boc-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine
Formel:
C₁₁H₁₇N₃O₂ Molecular Weight: 223,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059268 CAS-Nummer: 230301-11-8 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...