2-(Bromoacetyl)-6-methoxynaphthalene 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-6'-methoxy-2'-acetonaphthone
450060050
ACRO450060050EA
247
EUR
ACRO450060050
2-(Bromoacetyl)-6-methoxynaphthalene 96%
2-(Bromoacetyl)-6-methoxynaphthalene
Formel:
C₁₃H₁₁BrO₂ Molecular Weight: 279,13 g/mol Schmelzpunkt: 112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03964189 CAS-Nummer: 10262-65-4 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 110 to 114 °C |
HPLC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...