3'-Methoxybiphenyl-4-sulphonyl chloride 95%

Lieferant: Thermo Scientific
Gefahr

450480050
ACRO450480050EA 314 EUR
ACRO450480050
3'-Methoxybiphenyl-4-sulphonyl chloride 95%
3'-Methoxybiphenyl-4-sulphonyl chloride
Formel: C₁₃H₁₁ClO₃S
Molecular Weight: 282,74 g/mol
Schmelzpunkt: 81…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06409270
CAS-Nummer: 186550-26-5
UN: 3261
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR