Kaliumhydrogencarbonat, reinst

Lieferant: Thermo Fisher Scientific

Synonyme: Kaliumbicarbonat

450640010 450640050 450640250
ACRO450640010EA 43.5 EUR
ACRO450640010 ACRO450640050 ACRO450640250
Kaliumhydrogencarbonat, reinst
Kaliumhydrogencarbonat
Formel: KHCO₃
Molecular Weight: 100.12 g/mol
Schmelzpunkt: 292 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011402
CAS-Nummer: 298-14-6
Merck Index: 13,07691

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
Titration with HCl ≥98.5 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR