D(+)-Glucose Monohydrat 99+%, reinst

Lieferant: Thermo Fisher Scientific

Synonyme: Dextrose Monohydrat, Traubenzucker Monohydrat

450740010 450740050 450740250
ACRO450740010EA 34.7 EUR
ACRO450740010 ACRO450740050 ACRO450740250
D(+)-Glucose Monohydrat 99+%, reinst
D(+)-Glucose Monohydrat
Formel: C₆H₁₂O₆·H₂O
Schmelzpunkt: 83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149450
CAS-Nummer: 14431-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Water 8 to 10 %
HPLC ≥99.0 % (ELSD)
Residue after ign. (sulfated) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR