(4-Iodphenyl)essigsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

450900010 450900050
ACRO450900010EA 49.7 EUR
ACRO450900010 ACRO450900050
(4-Iodphenyl)essigsäure 97%
(4-Iodphenyl)essigsäure
Formel: (C₆H₄I)CH₂COOH
Schmelzpunkt: 140…141,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082985
CAS-Nummer: 1798-06-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to light brown Powder or solid
Infrared spectrum Conforms
Melting point 137 to 143 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR