3,4-Dimethylthiophen 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
450950010
450950050
ACRO450950010EA
122
EUR
ACRO450950010
ACRO450950050
3,4-Dimethylthiophen 97%
3,4-Dimethylthiophene
Formel:
C₆H₈S Molecular Weight: 112.1900 g/mol Siedepunkt: 144 °C (760 mmHg) Flammpunkt: 26 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01764731 CAS-Nummer: 632-15-5 UN: 1993 ADR: 3 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow (may darken on storage) Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...