2,4-Difluor-3-nitrobenzonitril 97%

Lieferant: Thermo Scientific
Gefahr

451420010
ACRO451420010EA 96.9 EUR
ACRO451420010
2,4-Difluor-3-nitrobenzonitril 97%
2,4-Difluor-3-nitrobenzonitril
Formel: C₇H₂F₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11226536
CAS-Nummer: 1186194-75-1
UN: 3276
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR