3-Pyridinsulfonylchloridhydrochlorid 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
451480050
ACRO451480050EA
198
EUR
ACRO451480050
3-Pyridinsulfonylchloridhydrochlorid 95%
Pyridine-3-sulphonyl chloride hydrochloride
Formel:
C5H5Cl2NO2S Schmelzpunkt: 144…145 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04035552 CAS-Nummer: 42899-76-3 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to yellow Powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % as diethylsulfonamide |
Elemental Analysis | 6.15 to 6.94 % (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...