L-Alanyl-L-glutamine
Lieferant: Thermo Fisher Scientific
451610010
451610050
ACRO451610010EA
42
EUR
ACRO451610010
ACRO451610050
L-Alanyl-L-glutamine
Proteine und Peptide
- Infrared spectrum: Conforms
- Loss on drying: ≤1%
- Residue after ignition: ≤0,1%
- Specific optical rotation: 8,0 to 12,0° (20 °C, 589 nm) (C=5, H₂O)
- Heavy metals (as Pb): ≤10 ppm
- Melting point: 213 to 220 °C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...