5-Pyrimidinboronsäure 97% may contain varying amounts of anhydride

Lieferant: Thermo Fisher Scientific
Achtung

451700250 451701000
ACRO451700250EA 262 EUR
ACRO451700250 ACRO451701000
5-Pyrimidinboronsäure 97% may contain varying amounts of anhydride
5-Pyrimidineboronic acid
Formel: C₄H₅BN₂O₂
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03002366
CAS-Nummer: 109299-78-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR