5-Pyrimidinboronsäure 97% may contain varying amounts of anhydride
Lieferant: Thermo Fisher Scientific
|
Achtung
|
451700250
451701000
ACRO451700250EA
262
EUR
ACRO451700250
ACRO451701000
5-Pyrimidinboronsäure 97% may contain varying amounts of anhydride
5-Pyrimidineboronic acid
Formel:
C₄H₅BN₂O₂ Schmelzpunkt: >300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03002366 CAS-Nummer: 109299-78-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...