1,2-Benzisoxazol-3-amin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
451710050
ACRO451710050EA
144
EUR
ACRO451710050
1,2-Benzisoxazol-3-amin 97%
1,2-Benzisoxazol-3-amin
Formel:
C₇H₆N₂O Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407353 CAS-Nummer: 36216-80-5 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Solid |
Infrared spectrum | Conforms |
Melting point | 107 to 112 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...