Imidazo[1,2-a]pyridin-6-carbonsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
451740010
451740100
ACRO451740010EA
55.3
EUR
ACRO451740010
ACRO451740100
Imidazo[1,2-a]pyridin-6-carbonsäure 97%
Imidazo[1,2-a]pyridin-6-carbonsäure
Formel:
C₈H₆N₂O₂ Schmelzpunkt: 250 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021498 CAS-Nummer: 139022-25-6 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...