L-Alanin 99%, meetSpec_Ph.Eur.

Lieferant: Thermo Fisher Scientific

Synonyme: H-L-Ala-OH, (S)-(+)-2-Aminopropionic acid, L(+)-2-Aminopropionic acid, A, (S)-(+)-Alanin, α-Alanin, H-Ala-OH, (S)-(+)-2-Aminopropionsäure, L(+)-Alanin, (S)-(+)-2-Aminopropansäure, L-α-Alanin

451850010 451850025 451852500
ACRO451850010EA 155 EUR
ACRO451850010 ACRO451850025 ACRO451852500
L-Alanin 99%, meetSpec_Ph.Eur.
L-Alanin
Formel: C₃H₇NO₂
Molecular Weight: 89.09 g/mol
Siedepunkt: 189
Schmelzpunkt: 314…316 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064410
CAS-Nummer: 56-41-7
Merck Index: 13,00203

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white to almost white Crystals or crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Sulfated ash ≤0.1 % (1 g, 600°C)
Appearance of solution Clear and not more intensely colored than reference solution BY6
Specific optical rotation +13.5° to +15.5° (20°C, 589 nm) (c=10, 6 N HCl) (dried substance)
Titration with HClO4 98.5 to 101.0 % (dried substance)
Sulfates ≤300 ppm
Chlorides ≤200 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Ninhydrin-positive substances ≤0.5 % (Total)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR