(2-Fluoropyridin-4-yl)methanol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(2-fluoro-pyridin-4-yl)-methanol, 2-Fluoro-4-hydroxymethylpyridine
452370010
452370050
ACRO452370010EA
124
EUR
ACRO452370010
ACRO452370050
(2-Fluoropyridin-4-yl)methanol 97%
(2-Fluoropyridin-4-yl)methanol
Formel:
C₆H₆FNO Molecular Weight: 127.12 g/mol Schmelzpunkt: 61…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200782 CAS-Nummer: 131747-60-9 |
Spezifikation Testergebnisse
Appearance | White to off-white to pink Powder |
Infrared spectrum | Conforms |
Melting point | 61 to 67 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...