3-Fluor-2-nitropyridin 98%

Lieferant: Thermo Scientific
Achtung

452450050
ACRO452450050EA 466 EUR
ACRO452450050
3-Fluor-2-nitropyridin 98%
3-Fluoro-2-nitropyridine
Formel: C₅H₃FN₂O₂
Schmelzpunkt: 33…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114127
CAS-Nummer: 54231-35-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR