5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(benzofuran-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Benzofuran, 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran
452630010
ACRO452630010EA
125
EUR
ACRO452630010
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan 97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Formel:
C₁₄H₁₇BO₃ Molecular Weight: 244,1 g/mol Schmelzpunkt: 72…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115374 CAS-Nummer: 519054-55-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow Solid |
Infrared spectrum | Conforms |
Melting point | 68 to 74 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...