L-Cysteic acid monohydrate 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Amino-3-sulphopropionic acid monohydrate

452840100
ACRO452840100EA 97.1 EUR
ACRO452840100
L-Cysteic acid monohydrate 99%
L-Cysteic acid monohydrate
Formel: C₃H₇NO₅S·H₂O
Schmelzpunkt: 253…254,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149544
CAS-Nummer: 23537-25-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Assay ≥98.5 %
Water ≤10 %
Specific optical rotation +7° to +8° (20°C, 589 nm) c=5 in water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR