Tetrahydropyran-2-ylmethylamine 97%

Lieferant: Thermo Fisher Scientific
Achtung

453370010 453370050
ACRO453370010EA 154 EUR
ACRO453370010 ACRO453370050
Tetrahydropyran-2-ylmethylamine 97%
Tetrahydropyran-2-ylmethylamine
Formel: C₆H₁₃NO
Siedepunkt: 110 °C (25 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD06738971
CAS-Nummer: 6628-83-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR