5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: tert-Butyl 2-bromo-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, tert-Butyl 2-bromo-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate

453590050
ACRO453590050EA 1008 EUR
ACRO453590050
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine 97%
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine
Formel: C₁₁H₁₅BrN₂O₂S
Molecular Weight: 319,23 g/mol
Schmelzpunkt: 94…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09951958
CAS-Nummer: 365996-06-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR