1-(2-Chloroethyl)-2-pyrrolidone 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2-chloroethyl)pyrrolidin-2-one, N-(beta-chloroethyl)-pyrrolidinone, 2-Pyrrolidinone, 1-(2-chloroethyl)-

453760010 453760050
ACRO453760010EA 74.7 EUR
ACRO453760010 ACRO453760050
1-(2-Chloroethyl)-2-pyrrolidone 95%
1-(2-Chloroethyl)-2-pyrrolidone
Formel: C₆H₁₀ClNO
Molecular Weight: 147.6 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00169064
CAS-Nummer: 51333-90-5

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR