2,5-Dichlorpyrimidin 95%

Lieferant: Thermo Scientific
Achtung

454040010
ACRO454040010EA 84 EUR
ACRO454040010
2,5-Dichlorpyrimidin 95%
2,5-Dichloropyrimidine
Formel: C₄H₂Cl₂N₂
Molecular Weight: 148,97 g/mol
Schmelzpunkt: 52…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00673131
CAS-Nummer: 22536-67-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or low melting solid
Infrared spectrum Conforms
Melting point 49 to 56 °C
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR