2-Quinoxalinol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
454460250
ACRO454460250EA
41.8
EUR
ACRO454460250
2-Quinoxalinol 98%
2-Quinoxalinol
Formel:
C₈H₆N₂O Schmelzpunkt: 271…272 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006722 CAS-Nummer: 1196-57-2 |
Spezifikation Testergebnisse
Appearance | White to yellow to tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 269 to 273 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...