Isoquinoline-3-carboxylic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
454490010
ACRO454490010EA
102
EUR
ACRO454490010
Isoquinoline-3-carboxylic acid 97%
Isoquinoline-3-carboxylic acid
Formel:
C₁₀H₇NO₂ Molecular Weight: 173,1681 g/mol Schmelzpunkt: –170…171,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075137 CAS-Nummer: 6624-49-3 |
Spezifikation Testergebnisse
Appearance | White to light brown Solid |
Infrared spectrum | Conforms |
Water | ≤10 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...