5-Thiazolcarbonsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
454540010
454540050
ACRO454540010EA
124
EUR
ACRO454540010
ACRO454540050
5-Thiazolcarbonsäure 98%
5-Thiazolecarboxylic acid
Formel:
C₄H₃NO₂S Molecular Weight: 129.1371 g/mol Schmelzpunkt: 208…210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03428539 CAS-Nummer: 14527-41-4 |
Spezifikation Testergebnisse
Appearance | White to yellow to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...