Ethyl-2-brom-1,3-thiazol-4-carboxylat 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-bromothiazole-4-carboxylate

454610010
ACRO454610010EA 72.1 EUR
ACRO454610010
Ethyl-2-brom-1,3-thiazol-4-carboxylat 96%
Ethyl-2-bromothiazole-4-carboxylate
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,08 g/mol
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788564
CAS-Nummer: 100367-77-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light orange to light brown Powder
Infrared spectrum Conforms
Melting point 66 to 72 °C
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR