2-Fluor-α-methyl-4-biphenylessigsäure 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

454660250 454660010 454660050
ACRO454660250EA 263 EUR
ACRO454660250 ACRO454660010 ACRO454660050
2-Fluor-α-methyl-4-biphenylessigsäure 98%
2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
Molecular Weight: 244.265 g/mol
Schmelzpunkt: 110…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079303
CAS-Nummer: 5104-49-4
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 110 to 116 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR