1-Boc-4-(methylamino)piperidin 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Methylamino)piperidine, N1-BOC protected, tert-Butyl 4-(methylamino)piperidine-1-carboxylate

454800010
ACRO454800010EA 56.5 EUR
ACRO454800010
1-Boc-4-(methylamino)piperidin 95%
1-Boc-4-(methylamino)piperidin
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,3046 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02259411
CAS-Nummer: 147539-41-1

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR