2-(Trifluoromethyl)pyrrolidine 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Aza-2-(trifluoromethyl)cyclopentane, 2-(Trifluoromethyl)tetrahydropyrrole
454810050
ACRO454810050EA
542
EUR
ACRO454810050
2-(Trifluoromethyl)pyrrolidine 97%
2-(Trifluoromethyl)pyrrolidine
Formel:
C₅H₈F₃N Molecular Weight: 139,12 g/mol Siedepunkt: 110…112 °C Dichte: 1,21 g/cm³ Flammpunkt: 26 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02663405 CAS-Nummer: 109074-67-1 UN: 1993 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to orange to light brown Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...