Cefotaxim Natriumsalz 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
454950010
454950050
454950250
ACRO454950010EA
0
EUR
ACRO454950010
ACRO454950050
ACRO454950250
Cefotaxim Natriumsalz 95%
Cefotaxime sodium salt
Formel:
C₁₆H₁₇N₅O₇S₂ Schmelzpunkt: 201…210 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00079073 CAS-Nummer: 64485-93-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤3.0 % (105°C, 8 hrs) |
HPLC | ≥94 % |
pH | 4.5 to 6.5 |
Specific optical rotation | +58° to +65° (20°C, 589 nm) (c=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...