2'-Desoxy-2'-fluoruridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

455000010
ACRO455000010EA 36 EUR
ACRO455000010
2'-Desoxy-2'-fluoruridin 97%
2'-Deoxy-2'-fluorouridine
Formel: C₉H₁₁FN₂O₅
Schmelzpunkt: 151 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01317293
CAS-Nummer: 784-71-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 153 °C
Loss on drying ≤1 % (1 g, 105 °C, 3 h)
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR