Amoxicillin 96%

Lieferant: Thermo Fisher Scientific

Synonyme: (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

455140050 455140250 455140010
ACRO455140050EA 0 EUR
ACRO455140050 ACRO455140250 ACRO455140010
Amoxicillin 96%
Amoxicillin

White to off-white Crystalline powder

Formel: C₁₆H₁₉N₃O₅S
Molecular Weight: 365.40 g/mol
Schmelzpunkt: 330 °C
MDL: MFCD00056860
CAS-Nummer: 26787-78-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water 10 to 15 %
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR