Amikacin disulphate
Lieferant: Thermo Scientific
Synonyme:
(S)-4-Amino-N-((1R,2S,3S,4R,5S)-5-amino-2-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4-(((2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxycyclohexyl)-2-hydroxybutanamide bis(sulphate)
455190050
ACRO455190050EA
91.9
EUR
ACRO455190050
Amikacin disulphate
Amikacin disulphate
Formel:
C₂₂H₄₃N₅O₁₃·2H₂SO₄ Molecular Weight: 781,77 g/mol Schmelzpunkt: 220…230 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00167475 CAS-Nummer: 39831-55-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤13 % (110°C, 3 h, 5 mm Hg) |
pH | 2 to 4 (1 % in water) |
Titration with NaOH | ≥87 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...