Amodiaquine dihydrochloride dihydrate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Amodiaquin dihydrochloride dihydrate, 4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 4-([7-Chloro-4-quinolinyl]amino)-2-([diethylamino]methyl)phenol, 4-[(7-Chloroquinolin-4-yl)amino]-2-[(diethylamino)methyl]phenol dihydrochloride

455200250
ACRO455200250EA 143 EUR
ACRO455200250
Amodiaquine dihydrochloride dihydrate
Amodiaquine dihydrochloride dihydrate
Formel: C20H22ClN3O .2HCl .2H2O
Molecular Weight: 464.812 g/mol (392.33 anhy)
Siedepunkt: 478 °C (760 mmHg)
Schmelzpunkt: 166…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078857
CAS-Nummer: 6398-98-7

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
Water 7.0 to 9.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR