Cefazolin Natriumsalz 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
455210010
455210050
ACRO455210010EA
85.2
EUR
ACRO455210010
ACRO455210050
Cefazolin Natriumsalz 98%
Cefazolin sodium salt
Formel:
C₁₄H₁₄N₈NaO₄S₃ Schmelzpunkt: 170…177 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00056883 CAS-Nummer: 27164-46-1 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 170 to 177 °C |
Water | ≤6 % |
HPLC | ≥97.5 % |
Specific optical rotation | -24° to -10° (20 °C, 589 nm) (c=1,H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...