Clarithromycin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-12,13-dihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-7-methoxy-3,5,7,9,11,13-hexamethyloxacyclotetradecane-2,10-dione

455220010
ACRO455220010EA 255 EUR
ACRO455220010
Clarithromycin 97%
Clarithromycin
Formel: C₃₈H₆₉NO₁₃
Molecular Weight: 747,96 g/mol
Schmelzpunkt: 222…231 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00865140
CAS-Nummer: 81103-11-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 222 to 231 °C
Water ≤2 %
HPLC ≥96.0 %
Specific optical rotation -87° to -97° (20 °C, 589 nm) (c=1,CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR