Imipenem monohydrate 98%

Lieferant: Thermo Fisher Scientific

455320010 455322500
ACRO455320010EA 618 EUR
ACRO455320010 ACRO455322500
Imipenem monohydrate 98%
Imipenem monohydrate
Formel: C₁₂H₁₇N₃O₄S
Schmelzpunkt: 193…198 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD09753321
CAS-Nummer: 74431-23-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Water 5 to 8 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR