6-(Hydroxymethyl)indol 97%

Lieferant: Thermo Fisher Scientific
Achtung

455540010 455542500
ACRO455540010EA 248 EUR
ACRO455540010 ACRO455542500
6-(Hydroxymethyl)indol 97%
6-(Hydroxymethyl)indole
Formel: C₉H₉NO
Schmelzpunkt: 45…50 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179595
CAS-Nummer: 1075-26-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 60 to 67 °C
Water ≤1 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR