Rifampicin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5,6,9,17,19,21-Hexahydroxy-23-methoxy- 2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl- 1-piperazinyl)formimidoyl]-2,7-(epoxypentadeca- [1,11,13]trienimino)-naphtho[2,1-b]furan- 1,11(2H)-dion-21-acetat, Rifampin

455620010 455620050 455620250
ACRO455620010EA 35.3 EUR
ACRO455620010 ACRO455620050 ACRO455620250
Rifampicin 95%
Rifampicin
Formel: C₄₃H₅₈N₄O₁₂
Schmelzpunkt: 183 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151389
CAS-Nummer: 13292-46-1

Bestellen

Spezifikation Testergebnisse

Appearance Light orange to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (100 mg, 60°C, 4 h) (vacuum)
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR