Imidazo[1,2-b]pyridazin 98%

Lieferant: Thermo Fisher Scientific
Achtung

455660010 455660050
ACRO455660010EA 0 EUR
ACRO455660010 ACRO455660050
Imidazo[1,2-b]pyridazin 98%
Imidazo[1,2-b]pyridazin
Formel: C₆H₅N₃
Molecular Weight: 119.12 g/mol
Schmelzpunkt: 54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07782103
CAS-Nummer: 766-55-2

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light grey Solid
Infrared spectrum Conforms
Melting point 50 to 57 °C
Water ≤3 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR